Sussman 29063C Replacment Heating Element, 6 kW, 240 VAC

Representative image only.
Sussman Mfr # 29063C – Item # EW-07603-21

$696.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Compatible WithSussman Steam Generator with 6 kW, 240 VAC power
  • Power (VAC)240
  • DescriptionReplacment Heating Element, 6 kW, 240 VAC

More About this Item

Keep your steam generator running smoothly with these parts and accessories.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat