Cole-Parmer Glass Capillary Tubes, Open Both Ends; 100/PK
Representative image only.
Cole-Parmer Item # EW-03013-63
Accessories for your melting point apparatus
  • Constructed of soda glass and easily sealed in a Bunsen flame
  • Supplied in a durable tube holder for added protection
  • Tubes are open on both ends
  • Previous part number SMP1/4
PRODUCT OPTIONS
$24.50USD / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass
  • Compatible With03011-40, 03011-41, 03011-45, 03011-47, 03011-49, 03013-60, 03013-61
  • Length (mm)100
  • Diameter (mm)1.9
  • DescriptionGlass Capillary Tubes, Open Both Ends; 100/PK
Key Features
  • Constructed of soda glass and easily sealed in a Bunsen flame
  • Supplied in a durable tube holder for added protection
  • Tubes are open on both ends
  • Previous part number SMP1/4
More About this Item
Capillary tubes measure 100 mm L, 1.9 mm OD, 1.3 mm ID, and 0.3 mm wall thickness.