Stir Rod, PTFE, 10 inch Length, Pack of 6

Representative image only.
Item # EW-06368-23
Useful for mixing chemicals and liquids
  • Constructed of chemically inert PTFE
  • Stir rod is 5/16” diameter by 10” length

SPECS & DESCRIPTION

Specifications & Description

  • MaterialPTFE
  • SterileYes
  • DescriptionStir Rod, Sterile, PTFE, 10 inch Length

Key Features

  • Constructed of chemically inert PTFE
  • Stir rod is 5/16” diameter by 10” length

More About this Item

These PTFE stir rods are ideally for stirring chemically aggressive liquids and come sterilized to avoid contamination.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat