Disposable Reagent Reservoir, Polystyrene, Sterile, 55 mL; 100/Pk

Representative image only.
Item # EW-24810-35
“V” shaped well design allows maximum reagent recovery
  • Easy to read graduation markings
  • Drip free pour-off spouts at every corner for spill free pouring
  • 1 year warranty

$190.00USD / Pkg of 100

SPECS & DESCRIPTION

Specifications & Description

  • Width (InchShort)1 1/10
  • SterileYes
  • Height (in)1 2/5
  • Max Volume (mL)55
  • Graduation Interval25- and 50-mL
  • Overall Length (in)4.5
  • Overall Length (mm)115
  • Wetted MatterialPolystyrene
  • DescriptionDisposable Reagent Reservoir, Polystyrene, Sterile, 55 mL; 50/Pk
  • Warranty1 year
1 Year Warranty

Key Features

  • Easy to read graduation markings
  • Drip free pour-off spouts at every corner for spill free pouring
  • 1 year warranty

More About this Item

Sterile reagent reservoirs are constructed of white polystyrene, providing easy to read graduation markings located on the top edge with a fill line on inner walls to indicate volume levels. The V-shaped bottom allows for efficient and nearly complete sample withdrawal.

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat