Starrett 660 Magnetic Base Indicator Holder with Triple Jointed Arm
Representative image only.
Starrett Mfr # 660 – Item # EW-09925-37
  • Articulating arm provides full 360° horizontal positioning and over 180° vertical positioning
  • Compact and versatile with three adjustable pivots controlled by a single knob
  • Will hold any indicator with a 3/8" stem or standard dovetail mount
  • Powerful magnetic base with 70-lb holding force
  • On/off switch
  • Dimensions: 1 3/16" x 1 9/16" x 1 3/8" (30 x 40 x 35 mm)
  • 1 year warranty
$174.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionMagnetic Base Indicator Holder with Triple Jointed Arm
  • Warranty1 year
1 Year Warranty
Key Features
  • Articulating arm provides full 360° horizontal positioning and over 180° vertical positioning
  • Compact and versatile with three adjustable pivots controlled by a single knob
  • Will hold any indicator with a 3/8" stem or standard dovetail mount
  • Powerful magnetic base with 70-lb holding force
  • On/off switch
  • Dimensions: 1 3/16" x 1 9/16" x 1 3/8" (30 x 40 x 35 mm)