Stainless steel spray gun (Stainless Steel) (0.25 in NPT(F))
Representative image only.
Mfr # YG4060SS – Item # EW-98515-15
No splashing
  • Recirculating gun has a contamination free fluid path to eliminate bacterial growth
  • Highly resistant to corrosive substances and fumes
  • Spray guns have a 1 GPM flow rate; max operating pressure of 75 psi
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • Connection TypeNPT(F)
  • Inlet Port1/4" NPT(F)
  • Max Pressure (PSI)4050
  • Max Temperature (° F)320
  • Max Temperature (° C)150
  • Wetted MaterialsStainless Steel
  • DescriptionStainless steel spray gun (Stainless Steel) (0.25 in NPT(F))
Key Features
  • Recirculating gun has a contamination free fluid path to eliminate bacterial growth
  • Highly resistant to corrosive substances and fumes
  • Spray guns have a 1 GPM flow rate; max operating pressure of 75 psi
More About this Item
These Masterflex® Tank Dispensing Spray Guns feature a special tip to eliminate splashing. Choose from PTFE or stainless steel construction for a wide range of applications.