Spirax Sarco Cast iron inverted bucket steam trap; 3/4" NPT(F), 15 psig
Representative image only.
Spirax Sarco Mfr # B1H-15 3/4" 64159 – Item # EW-07600-62
Keep steam space free of condensate to provide maximum system efficiency
  • All stainless steel working parts provide good resistance to corrosion and wear
  • Inverted bucket steam traps rely on the difference in density between steam and condensate to drain condensate as it forms.
  • Condensate is discharged close to steam temperature
PRODUCT OPTIONS
$243.10USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))0.75
  • Outlet Port3/4" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port3/4" NPT(F)
  • Max Pressure (PSI)15
  • Max Temperature (° F)450
  • Max Temperature (° C)232
  • Wetted MaterialsCast iron
  • DescriptionCast iron inverted bucket steam trap; 3/4" NPT(F), 15 psig
Key Features
  • All stainless steel working parts provide good resistance to corrosion and wear
  • Inverted bucket steam traps rely on the difference in density between steam and condensate to drain condensate as it forms.
  • Condensate is discharged close to steam temperature
More About this Item
Typical applications:
  • Steam main drip stations
  • Laundry equipment
  • Industrial dryers
  • Storage tanks