Cole-Parmer Spex® 54006 Diffuser, for Liquid Nitrogen, 3/8" NPT(F)
Representative image only.
Cole-Parmer Item # EW-04591-64
Accessories for Freezer/Mill® cryogenic grinders to simplify your processes
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Use with transfer hoses 04500-54 or 04576-46 to manually fill liquid nitrogen
PRODUCT OPTIONS
$273.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Thread size NPT(F) (port 1) (in)3/8
  • DescriptionDiffuser, for Liquid Nitrogen, 3/8\" NPT(F)
Key Features
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Use with transfer hoses 04500-54 or 04576-46 to manually fill liquid nitrogen
More About this Item
Compatible with Freezer/Mill® cryogenic grinders 04500-39 to -42, 04577-77, and 04577-92 to -94.