Cole-Parmer Spex® 3527IM 40 mm Bubble Free Insert for 04576-05; 1000/PK
Representative image only.
Cole-Parmer Item # EW-04577-24
Enhance the performance of your XRF X-Cell sample cup
  • Compatible with 40 mm closed X-Cell
  • Compatible with normal optics XRF spectrometers
  • Forms inverted-cone shelf
PRODUCT OPTIONS
$1,230.00USD / Pkg of 1000
BUY MORE & SAVE
  • 1-4$1,230.00USD / Pkg of 1000
  • 5+$984.00USD / Pkg of 1000
SPECS & DESCRIPTION
Specifications & Description
  • Tube MaterialPolyethylene
  • Description40 mm Bubble Free Insert for 04576-05; 1000/PK
Made in the U.S.A.
Key Features
  • Compatible with 40 mm closed X-Cell
  • Compatible with normal optics XRF spectrometers
  • Forms inverted-cone shelf
More About this Item
Use the bubble-free insert with the 3527 04576-05 40 mm XRF liquid cell to prevent trapped air from forming a bubble under the window film when the cell is upright. The insert is pushed into the cell before the sample is added and forms an inverted-cone shelf under which air bubbles can be trapped once the cell is filled and sealed. To use, simply push the insert into the X-Cell after it is partially filled. It is designed to fit into the major suppliers of normal optics XRF spectrometers made by Diano, Rigaku, and Shimadzu.