Spex Protects Presaturated Surface Cleaning Wipes, 70% IPA / 30% DI; 100/Canister

Representative image only.
Spex Protects Item # EW-78901-42
Strong, nonwoven polypropylene-viscose blend cloth is soft and lint-free ─ will not scratch surfaces
  • Contains 70% isopropyl alcohol and 30% DI water
  • Safe to use on electronics, keyboards, and touchscreens
  • Canister contains 100 premoistened 6" x 7 1/2" wipes
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Formatpolypropylene-viscose blend
  • Width (InchShort)6 1/2
  • Height (in)7
  • ApplicationsElectronics, tools, workspaces
  • DescriptionSurface Cleaning Wipes, 70% IPA / 30% DI; 100/Canister
  • Warranty1 year
1 Year Warranty

Key Features

  • Contains 70% isopropyl alcohol and 30% DI water
  • Safe to use on electronics, keyboards, and touchscreens
  • Canister contains 100 premoistened 6" x 7 1/2" wipes
  • 1 year warranty

More About this Item

The 100% polypropylene with honeycomb pattern means a durable yet soft wipe, great for removing dirt and grime from hard, nonporous surfaces. Wipes are premoistened with a blend of pure, reagent-grade 70% isopropyl alcohol and 30% reagent-grade deionized water (no surfactants) to minimize residue after cleaning.


Available for sale in the United States only. Does not ship to California.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat