Spex CertiPrep RSCN9C-2X Cyanide, Complex Certified Reference Standard, 1,000 µg/mL in 2% KOH; 500 mL

Representative image only.
Spex CertiPrep Item # EW-04569-42
Reference standard for ion chromatography
  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
  • 1,000 ppm complex cyanide reference standard in 2% KOH matrix
  • 500 mL bottle

$157.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Concentration1,000 µg/mL (1,000 ppm)
  • Matrix2% KOH
  • Element/IonCyanide, complex
  • FormLiquid
  • Volume (mL)500
  • DOT InformationHazardous; Potassium hydroxide solution; Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolCorrosion
  • UN NumberUN1814
  • Shelf Life (months)6
  • DescriptionCyanide, Complex Certified Reference Standard, 1,000 µg/mL in 2% KOH; 500 mL

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
  • 1,000 ppm complex cyanide reference standard in 2% KOH matrix
  • 500 mL bottle

More About this Item

Ion chromatography (IC) is an analytical process for the separation of ions based on charge affinity. IC can be used for a variety of different kinds of charged analytes from single elements to large proteins. In order to ensure accurate analysis, quality standards which are traceable and stable are necessary. Spex CertiPrep® IC standards are of the highest quality available for the analytical laboratory.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Hazardous material shipping fees may apply.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat