Spectra Por S/P 3 3 Dialysis Membrane Disc, 3500 to 3500 MWCO, 47 mm dia; pack of 50.
Representative image only.
Spectra Por Mfr # 132486 – Item # EW-02890-67
Available in a variety of dimensions to meet your application needs
  • Regenerated Cellulose (RC) membrane for the gentle separation and purification of solutes
  • 1 year warranty
PRODUCT OPTIONS
$265.00USD / Pkg of 50
SPECS & DESCRIPTION
Specifications & Description
  • MaterialRegenerated Cellulose
  • Max Molecular Weight Cut Off (Mwco) (Dalton)3500
  • Disc Diameter (mm)47
  • Description3 Dialysis Membrane Disc, 3500 to 3500 MWCO, 47 mm dia; pack of 50.
  • Warranty1 year
1 Year Warranty
Key Features
  • Regenerated Cellulose (RC) membrane for the gentle separation and purification of solutes
  • 1 year warranty
More About this Item
MWCO of 3500 Dalton