Spectra Por 132743 Weighted Tubing Closures; max gripping width, 23mm; 10/box
Representative image only.
Spectra Por Mfr # 132743 – Item # EW-02899-52
The most convenient method for sealing dialysis tubing without the risk of leaking
  • Easy to open and reseal
SPECS & DESCRIPTION
Specifications & Description
  • Tubing Sealing Width (mm)23
  • ColorWhite
  • DescriptionWeighted Tubing Closures; max gripping width, 23mm; 10/box
Key Features
  • Easy to open and reseal
More About this Item
Weighted closures feature a fluorocarbon-coated stainless steel bar. The weighted closure acts like an anchor to maintain a vertical orientation of the sample.