Spectra Por 131054T Biotech-Grade Dialysis Tubing Trial Kit, 0.1-0.5 kDalton, 16 mm
Representative image only.
Spectra Por Mfr # 131054T – Item # EW-02980-11
  • 1 year warranty
PRODUCT OPTIONS
$218.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialCellulose Ester
  • Diameter (mm)10
  • DescriptionBiotech-Grade Dialysis Tubing Trial Kit, 0.1-0.5 kDalton, 16 mm
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
These convenient kits contain everythig needed to evaluate dialysis membranes for your application. The small roll is also ideal for low volume projects.
Cellulose Ester (CE) offers a wider range of more precise MWCO’s for critical separations; packaged wet in 0.05% sodium azide.