Snyder 66108 Standard square stackable tank, 330 gallons

Representative image only.
Snyder Mfr # 66108 – Item # EW-06960-04
Stackable polyethylene IBC’s deliver durable, long-lasting performance
  • UN/DOT certified
  • Four-way forklift access
  • 2” bottom drain valve
  • 2 year warranty

PRODUCT OPTIONS

$1,771.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • MaterialPolyethylene
  • Wall thickness (in)0.5
  • Volume (Gal)330
  • Volume (Liters)1249
  • Width (in)45
  • Height (in)59
  • Height (cm)149.86
  • Length (in)45
  • Length (cm)114.3
  • Fill Port6 in
  • DescriptionStandard square stackable tank, 330 gallons
  • Warranty2 Years
Two year warranty

Key Features

  • UN/DOT certified
  • Four-way forklift access
  • 2” bottom drain valve
  • 2 year warranty

More About this Item

The plastic resin and processing techniques used with these IBC’s ensure the container wall thickness of ½" or more is cured optimally. The physical properties of the polyethylene are maximized to consistently deliver a durable and long-lasting product. The all-plastic construction of these tanks also resists the corrosion attack and associated maintenance costs common in metal or composite IBC constructions. These heavy-duty IBC’s actually weigh less than comparative material alternatives, which reduce a filled IBC's payload freight and handling expenses. IBC’s are DOT and UN approved to handle and transport hazardous liquids weighing up to 16 lbs. per gallon. Stack full tanks two high for transportation and three high for storage. All tanks have a 6” fill cap, except for model 06960-22 and -24, which have a 10” fill cap.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat