SHOWA 574-10 Glove; Size, Extra-Large; 12 Pairs
Representative image only.
SHOWA Mfr # 574-10 – Item # EW-86288-63
Protects against acids, caustics, and dyes
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
$240.00USD / Pkg of 12
BUY MORE & SAVE
  • 1-5$240.00USD / Pkg of 12
  • 6+$216.00USD / Pkg of 12
SPECS & DESCRIPTION
Specifications & Description
  • MaterialLatex
  • Thickness (mil)50
  • SizeX-Large
  • Length (in)23
  • Length (cm)58
  • ColorWhite
  • SterileNo
  • DescriptionGlove; size, extra-large; 12/pack
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
More About this Item
Tuff Guard™ latex gloves provide protection against acids, caustics, and dyes. Use for deep-tank plating and agricultural applications. Pack of 12 pairs.