Shimpo DT-326B Portable LED Stroboscope Kit

Representative image only.
Shimpo Mfr # DT-326B – Item # EW-53565-75
High-intensity light emitting diodes (LEDs) in a rugged all-metal design
  • Provides a long flash life - 5 times longer life then traditional xenon flashtubes
  • Ideal for motion and vibration analysis, and preventive maintenance programs

$625.50USD / Each
$695.00USD / Each

$150.00USD / Each

for an additional
$150.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Min Flash Range (fpm)60
  • Max Flash Range (fpm)120000
  • Flash Accuracy (fpm)0.02% ±1 digit at 77°F
  • External InputPhotocoupler input 12mA/DC12V
  • Display Type6-character backlit LCD
  • Battery Life80 min (Eco); 20 min (normal)
  • Length (in)8 45/64
  • Width (in)2 13/32
  • Length (cm)22
  • Width (cm)6.1
  • DescriptionPortable LED Stroboscope Kit
Two year warrantyNIST Traceable Calibration

Includes

100 to 240 VAC adapter, input/output connect kit, carrying case, and four AA batteries.

Key Features

  • Provides a long flash life - 5 times longer life then traditional xenon flashtubes
  • Ideal for motion and vibration analysis, and preventive maintenance programs

More About this Item

Easily monitor and inspect a variety of machine tools, components, raw or finished product, print media and many other applications under various speeds with this easy-to-use stroboscope. Unit features pulse control (adjustable pulse/focus) making it ideal for fine and gross viewing of fast-moving, fine movement details of small, high-speed moving objects like drill bits, fans and saw blades, micro motors, and more. The flash delay/phase shift control allows on-the-fly inspections of high-speed machinery and printing, without stopping the equipment, reducing critical downtime. Use the input/output connector port for internal or external LED flash triggering and for input external triggering sources such as remote sensors. Additional features include a simple manual dial, push button (X2), (/2), manual dial for flash rate controls, (+/–) control, and backlit LCD.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat