Sherwood Scientific 50035008 Fluid Bed Dryer Tub and Assembly, Non-Sealed, Stainless Steel, 5-Liter
Representative image only.
Sherwood Scientific Mfr # 50035008 – Item # EW-02658-54
Quickly and evenly dry particles as small as 5 μm
  • Faster and gentler than conventional drying cabinets—ideal for food products, polymers, soils, chemicals, pharmaceuticals, minerals, and waste materials
  • Variety of accessories provide easy customization for your drying applications
SPECS & DESCRIPTION
Specifications & Description
  • Materials of ConstructionStainless steel
  • DescriptionFluid Bed Dryer Tub and Assembly, Non-Sealed, Stainless Steel, 5-Liter
Key Features
  • Faster and gentler than conventional drying cabinets—ideal for food products, polymers, soils, chemicals, pharmaceuticals, minerals, and waste materials
  • Variety of accessories provide easy customization for your drying applications
More About this Item
Non-Sealed Tub Assemblies are available in glass or stainless steel and require a filter bag, sold separately below.
Model 02658-02 includes a port to add extra sample and a port for a temperature/humidity probe (sold separately).
Order fluid bed dryer, filter bags, and optional accessories separately for a complete fluid bed dryer system.
High-grade stainless steel tubs are for high product purity and good chemical and corrosion resistance at high temperatures. The 2- and 5-L units handle samples up to 11 lb (5.0 kg). Required filter bags sold separately.