Seward Stomacher® 400 Circulator Strainer Bags, Sterile, 80-400 mL; 200/PK
Representative image only.
Seward Mfr # BA6141/STR – Item # EW-36860-01
Eliminates the risk of crosss contamination
  • 1 year warranty
  • Manufactured from high-grade, food quality polythene
  • Strainer / Filter blender bags are for samples that produce a large amount of debris or fat or harder samples such as shellfish.
  • Strainer / Filter blender bags contain a robust strainer element to make it easier to pipette the sample produced (pore size 0.5mm, prevents pore blockage and non-homogenous sample).
  • Compatible with all 400 lab blenders homogenizers
  • 1 year warranty
PRODUCT OPTIONS
$356.25USD / Pkg of 200
Often bought with...
SPECS & DESCRIPTION
Specifications & Description
  • Bag TypePaddle Blender Bags
  • Bag Capacity (mL)400
  • Bag Capacity (L)0.4
  • DescriptionSterile Strainer/Filter Bags, 80-400 mL; 200/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
  • Manufactured from high-grade, food quality polythene
  • Strainer / Filter blender bags are for samples that produce a large amount of debris or fat or harder samples such as shellfish.
  • Strainer / Filter blender bags contain a robust strainer element to make it easier to pipette the sample produced (pore size 0.5mm, prevents pore blockage and non-homogenous sample).
  • Compatible with all 400 lab blenders homogenizers
  • 1 year warranty
More About this Item
Choose from a variety of bags to use with your lab blender. Blending bags are made from high quality food grade nontoxic virgin LDPE granule, extruded to 65µm film thickness. All blender bags are irradiated sterile. A certificate or irridiation is available upon request.
The Stomacher bag range has been designed to fulfil all applications from sample blending, straining and storage, including bags designed for extensive blending (processes requiring more than 15 minutes on the machine) and small samples for life science applications.