Sensorex TX2000 1/4-DIN pH/ORP Transmitter/Controller; 100-240 VAC

Representative image only.
Sensorex Mfr # TX2000 – Item # EW-19066-06
Intelligent analyzer for reliable pH and ORP measurements
  • Large, graphical LCD with auto-sense backlight and contrast can be easily viewed in all lighting conditions
  • Easy menu-driven operation with continuous diagnostic tests to ensure proper operation
  • Dual diaplay with measurement and temperature
  • Backup memory retains settings during power outage
  • Automatic temperature compensation
  • Up to 3-point calibration with USA or NIST buffers
  • Panel- or wall-mount NEMA 4X, IP65 rated housing
  • 1 year warranty

$1,263.00USD / Each

$164.50 - $237.00USD / Each

Time to recalibrate? Click on the calibration service to view details, then order.
Include Accessories Today!

SPECS & DESCRIPTION

Specifications & Description

  • Control TypeOn/Off
  • Display TypeLCD
  • Housing MaterialNEMA 4X (IP 65)
  • Min pH (pH)-2
  • Max pH (pH)16
  • pH Accuracy±1% + 1 digit
  • Min mV (mV)-1999
  • Max mV (mV)1999
  • Min Temperature (° C)-30
  • Max Temperature (° C)130
  • Min Temperature (° F)-22
  • Max Temperature (° F)266
  • Password ProtectionYes
  • Output Current4-20mA (x2)
  • Output Relay2 SPDT relays
  • Width (cm)9.53
  • Height (cm)9.53
  • Width (in)3 3/4
  • Height (in)3 3/4
  • Depth (in)5 3/16
  • Description1/4-DIN pH/ORP Transmitter/Controller; 100-240 VAC
  • Warranty1 year
1 Year WarrantyIP 65

Key Features

  • Large, graphical LCD with auto-sense backlight and contrast can be easily viewed in all lighting conditions
  • Easy menu-driven operation with continuous diagnostic tests to ensure proper operation
  • Dual diaplay with measurement and temperature
  • Backup memory retains settings during power outage
  • Automatic temperature compensation
  • Up to 3-point calibration with USA or NIST buffers
  • Panel- or wall-mount NEMA 4X, IP65 rated housing
  • 1 year warranty

More About this Item

This full-featured pH/ORP transmitter/controller is designed for drinking water, wastewater, and general-purpose applications. The analyzer offers a wide range of features: two adjustable high/low relays, one wash control, and two isolated 4 to 20 mA outputs (one for the selected measurement and one for temperature), and enhanced measurement stability with sample averaging functionality. Automatic calibration with preset standards for up to three buffers. Calibration data can be password protected with a programmable security code.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat