Scienceware 37911-1010 Pipette Pumps with Rapid-Release Lever, 10 mL, Green; Each
Representative image only.
Scienceware Mfr # 37911-1010 – Item # EW-06221-33
Easy, one-handed, operation!
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • Max Volume (µL)10000
  • Max Volume (mL)10
  • Max Dispensing (mL)10
  • Body MaterialHDPE
  • Color CodeGreen
  • DescriptionPipette Pumps with Rapid-Release Lever, 10 mL, Green; Each
More About this Item
Pipette Pumps feature a knurled wheel for precise level adjustment. Attach to pipette and dispense by depressing plunger or rotating wheel. Press the rapid-release lever for quick and easy dispensing. All models are color-coded. High-density polyethylene (HDPE) construction.