Weighing UnitsGram, kilogram, carat, pound, ounce, troy ounce, Hong Kong tael, Singapore tael, Taiwan tael, grain, pennyweights, milligram, parts per pound, China tael, mommes, Austrian carat, tola, baht, mesghal and Newton
DescriptionEntris® II Advanced Toploading Balance, 10,200 g x 0.1 g; 100 to 240 VAC, 50/60 Hz
Warranty2 Years
Key Features
Internal electronic level sensor provides built-in real-time level support
CalAuditTrail provide automatic documentation of calibration, leveling, and date-and-time stamps
Hybrid screen combines LED and graphic touch display to improve functionality
Optional isoCAL internal calibration saves you time and money for accurate weighing results
Parts offer a high chemical resistance so there is no need to worry over spills
Easily transfer data directly to a PC or USB stick with multiple connectivity options
Weighs up to 10,200 g│Offers external calibration
2 year warranty
More About this Item
Sartorius Entris® II Advanced precision toploading balances offer higher weighing capacities than their analytical counterparts. The units are constructed of chemical-resistant polybutylene terephthalate, stainless steel, and glass to provide enhanced durability. In addition to high capacity and durability, the balances have advanced features that include dosing, counting, percentage weighing, mixing, totalizing, animal weighing, free factor, density determination, statistics, peakhold, checkweighing, mass unit conversion, and pipette smart test. They enable multiple connectivity options to external devices including RS-232 and USB Type C which offers quick and easy data transfer to a USB stick, PC, or printer.
The Entris® II Advanced line offers enhanced features to ensure accuracy of your measurements. Built-in real-time level support with an internal electronic level sensor continuously monitors proper leveling, provides alarm messaging when your balance isn’t level, and offers interactive user guidance. Integrated protection systems feature three configurable levels to determine valid weighing data and ensure only valid data is transferred to external devices to enhance reliability and accuracy of results. CalAuditTrail provides constant automatic documentation of calibration and levelling processes, and date-and-time stamp.