Rubberfab Torque Tee Sanitary Clamp Securing Device, 3-Piece Extension Set, 1/4"
Representative image only.
Rubberfab Mfr # TR-EXT-.25-SET – Item # EW-23508-04
Ensure the security of sanitary connections with preset torque output
  • Works with any sanitary clamp wing nut and Rubberfab's Torque-Rite® nut
  • Easily reach into tight or awkward spaces
  • Choose the appropriate torque output per the clamp manufacturer's specifications
  • Audible click alerts you when the torque has been reached
  • Ratcheting mechanism disengages for easy clamp removal and is sealed within handle for durability
PRODUCT OPTIONS
$43.75USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Drive (in)0.25
  • DescriptionTorque Tee Sanitary Clamp Securing Device, 3-Piece Extension Set, 1/4"
Key Features
  • Works with any sanitary clamp wing nut and Rubberfab's Torque-Rite® nut
  • Easily reach into tight or awkward spaces
  • Choose the appropriate torque output per the clamp manufacturer's specifications
  • Audible click alerts you when the torque has been reached
  • Ratcheting mechanism disengages for easy clamp removal and is sealed within handle for durability
  • "T" shape creates effective leverage for fast, easy one-handed tightening
More About this Item
The Rubberfab Torque Tee and universal socket are designed to safely and securely install and remove sanitary clamps. Available with a family of components, the Torque Tee handle creates effective tightening leverage, even in hard-to-reach spaces. If you need to loosen or remove the clamp, simply disengage the ratcheting mechanism to lock the handle. The universal socket is required when using the Torque Tee.