Red Lion PAXT0010 Thermocouple/RTD Panel Meter; 11-26VDC/24VAC
Representative image only.
Red Lion Mfr # PAXT0010 – Item # EW-65590-02
Upgradeable for customized solutions – Choose from temperature, process, voltage, and current inputs
  • Process, Voltage, Current, Temperature, And Strain Gauge Inputs
  • 6-Digit 0.56'' Red Sunlight Readable Display
  • Variable-intensity display for viewing in all lighting conditions
  • 16 Point Scaling For Non-Linear Processprogrammable Function Keys/User Inputs
  • 9 Digit Totalizer (Integrator) With Batching
PRODUCT OPTIONS
$328.00USD / Each
sign in for your price.
$112.00USD / Each
for an additional
$112.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • DIN Size1/8
  • Control Input RTDYes
  • Control Input ThermocoupleYes
  • Display Type0.56" LED
  • Display Digits5
  • Max Power VAC (VAC)24
  • Min Power VAC (VAC)24
  • Max Power VDC (VDC)36
  • Min Power VDC (VDC)11
  • Max Operating Temperature (° F)122
  • Min Operating Temperature (° F)32
  • EnclosureNEMA 4X; IP65
  • DescriptionThermocouple/RTD Panel Meter; 11-26VDC/24VAC
Two year warranty
Key Features
  • Process, Voltage, Current, Temperature, And Strain Gauge Inputs
  • 6-Digit 0.56'' Red Sunlight Readable Display
  • Variable-intensity display for viewing in all lighting conditions
  • 16 Point Scaling For Non-Linear Processprogrammable Function Keys/User Inputs
  • 9 Digit Totalizer (Integrator) With Batching
  • Optional Custom Units Overlay W/Backlight
  • Our Setpoint Alarm Outputs (W/Option Card)
  • Communication And Bus Capabilities (W/Option Card)
  • Retransmitted Analog Output (W/Option Card)
  • Crimson Programming Software
  • NEMA 4X/IP65 Sealed Front Bezel
More About this Item
PART NUMBERS REPRESENT VARIOUS MODELS AND FEATURES. ORDER OUTPUT OR COMMUNICATIONS CARDS SEPARATELY
The PAX® Analog Panel Meters offer many features and performance capabilities to suit a wide range of industrial applications. Available in five different models to handle various analog inputs, including DC Voltage/Current, AC Voltage/Current, Process, Temperature, and Strain gauge Inputs. The optional plug-in output cards allow the opportunity to configure the meter for present applications, while providing easy upgrades for future needs. The meters employ a bright 0.56'' LED display. The unit is available with a red sunlight readable or a standard green LED. The intensity of display can be adjusted from dark room applications up to sunlight readable, making it ideal for viewing in bright light applications.
The meters provide a MAX and MIN reading memory with programmable capture time. The capture time is used to prevent detection of false max or min readings which may occur during start-up or unusual process events.
The signal totalizer (integrator) can be used to compute a time-input product. This can be used to provide a readout of totalized flow, calculate service intervals of motors or pumps, etc. The totalizer can also accumulate batch weighing operations.
The meters have four setpoint outputs, implemented on Plug-in option cards. The Plug-in cards provide dual FORM-C relays (5A), quad FORM-A (3A), or either quad sinking or quad sourcing open collector logic outputs. The setpoint alarms can be configured to suit a variety of control and alarm requirements.