Process, Voltage, Current, Temperature, And Strain Gauge Inputs
6-Digit 0.56'' Red Sunlight Readable Display
Variable-intensity display for viewing in all lighting conditions
16 Point Scaling For Non-Linear Processprogrammable Function Keys/User Inputs
9 Digit Totalizer (Integrator) With Batching
Optional Custom Units Overlay W/Backlight
Our Setpoint Alarm Outputs (W/Option Card)
Communication And Bus Capabilities (W/Option Card)
Retransmitted Analog Output (W/Option Card)
Crimson Programming Software
NEMA 4X/IP65 Sealed Front Bezel
2 year warranty
More About this Item
PART NUMBERS REPRESENT VARIOUS MODELS AND FEATURES. ORDER OUTPUT OR COMMUNICATIONS CARDS SEPARATELY
The PAX® Analog Panel Meters offer many features and performance capabilities to suit a wide range of industrial applications. Available in five different models to handle various analog inputs, including DC Voltage/Current, AC Voltage/Current, Process, Temperature, and Strain gauge Inputs. The optional plug-in output cards allow the opportunity to configure the meter for present applications, while providing easy upgrades for future needs. The meters employ a bright 0.56'' LED display. The unit is available with a red sunlight readable or a standard green LED. The intensity of display can be adjusted from dark room applications up to sunlight readable, making it ideal for viewing in bright light applications.
The meters provide a MAX and MIN reading memory with programmable capture time. The capture time is used to prevent detection of false max or min readings which may occur during start-up or unusual process events.
The signal totalizer (integrator) can be used to compute a time-input product. This can be used to provide a readout of totalized flow, calculate service intervals of motors or pumps, etc. The totalizer can also accumulate batch weighing operations.
The meters have four setpoint outputs, implemented on Plug-in option cards. The Plug-in cards provide dual FORM-C relays (5A), quad FORM-A (3A), or either quad sinking or quad sourcing open collector logic outputs. The setpoint alarms can be configured to suit a variety of control and alarm requirements.