Universal process inputs: DC current, DC voltage, process signals, thermocouples and RTDs
Universal power supply: 50 to 250 VAC or 21.6 to 250 VDC
Built-in USB programming port
Meter update rate up to 100/second
Easy PAX style programming
Setpoints; dual and quad relay, quad sinking and sourcing
Retransmitted analog output, 0 to 10 VDC or 0 (4) to 20 mA
MODBUS communication; RS-232, RS-485, DeviceNet™ and Profibus
NEMA 4X (IP65) faceplate/IP20 enclosure
1 year warranty
More About this Item
The PAX2A is the next generation in panel meters. From its dual-line display to its universal capability the meter is packed full of features that sets it apart from any other meter. The input, total, min, max or setpoints can be displayed on the 0.7" height 6- digit main LCD display. The main display also offers three programmable colors, red, orange and green. The color change can be tied to the setpoints that will provide the operator a visual display of changing conditions in the application. The second display line, 0.35" height 9-digit, green LCD can be programmed any of the above parameters as well. The 9-digit display allows for totalizing application that easily exceeds the normal 6-digit displays. In addition to the dual displays, the meter also offers a 3 digit programmable mnemonic, which allows you to add identification to the display value.
Meter accepts DC currents, DC voltages, process signals, thermocouples and RTD temperature sensors and offers a universal power input as well. Whether its 50 to 250 VAC or 21.6 to 250 VDC, just hook it up. The PAX2A provides the maximum in application flexibility allowing you to stock one meter for all the above applications.
The ideal application for the PAX2A is where two parameters need to be visualized at the same time, like reading a flow rate through a pipeline while totalizing the gallons or indicating the temperature of an oven while displaying the setpoint value.