Red Lion G307K200 Operator Interface with Color Display; 7"
Representative image only.
Red Lion Mfr # G307K200 – Item # EW-68515-97
Human machine interface that strikes the perfect balance of advanced features and value!
  • Unit's configuration is stored in nonvolatile Flash memory
  • NEMA 4/IP65 front panel
  • Resistive analog touch-screen
  • Bright TFT display with full color support
  • Both Kadet models are programmed with Red Lion's free Crimson® software
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Communication OutputSerial ports: one RS-232 port, two RS-232/422/485; one Ethernet port (10 Mbps); 115,200 baud each
  • Memory Capacity2MB of nonvolatile flash memory; CompactFlash Type II slot
  • Enclosure RatingNEMA 4 / IP65
  • Display Type5.6\", 320 x 234, 256-color, LED backlight
  • Power (VDC)12 to 24
  • DescriptionOperator Interface with Color Display; 7\"
  • Warranty2 Years
Two year warranty
Key Features
  • Unit's configuration is stored in nonvolatile Flash memory
  • NEMA 4/IP65 front panel
  • Resistive analog touch-screen
  • Bright TFT display with full color support
  • Both Kadet models are programmed with Red Lion's free Crimson® software
  • 2 year warranty
More About this Item
G3 Kadet with TFT Color Display
The Kadet is the perfect solution for applications that require the operator to monitor and control more than just a single device. With multiple serial ports and an Ethernet port, these Kadets can connect to multiple serial and Ethernet devices simultaneously, including PLCs, motor drives, bar code scanners, etc.
Programmed with Red Lion’s free Crimson 2.0 software.
The G307K2 - 7" display (3 serial and 1 ethernet port)
performs the functions of a multiple protocol converter. The Ethernet port supports up to four protocols simultaneously, allowing dissimilar Ethernet-based products to communicate with one another.