Red Lion DT800010 Adj Timebase Tach W/Red Backlighting
Representative image only.
Red Lion Mfr # DT800010 – Item # EW-68484-61
SPECS & DESCRIPTION
Specifications & Description
  • Display Type0.6\" backlit LCD
  • Display Digits5
  • Max Power VDC (VDC)28
  • Min Power VDC (VDC)9
  • EnclosureNEMA 4X; IP65
  • DescriptionAdj Timebase Tach W/Red Backlighting
More About this Item
The DT8 is a self-powered rate indicator which features selectabletimebase increments by setting the appropriate DIP switches on the rear of the unit. The internal 3.0 VDC lithium battery will operate continuously for at least 7 years. It has a 5-digit LCD display with 0.6 inch (15.2mm) high digits.
The unit is constructed of a lightweight, high impact plastic case with a clear viewing window. The sealed front panel meets NEMA 4X/IP65 specifications for wash-down and/or dusty environments, when properly installed. The optional Micro Line/Sensor Power Supply (RLMLPS10000) is designed to attach to the rear of an installed DT8. The optional supply can be powered from an 85 to 250 VAC source, and can provide power for the backlighting of a unit and most sensors.