Red Lion 308TX Ethrnet Swtch8port 10/100BaseTX Din-Rail
Representative image only.
Red Lion Mfr # 308TX – Item # EW-17782-96
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Number of Ports8
  • Max Input Voltage (VDC)30
  • Min Input Voltage (VDC)10
  • Max Input Current (mA)250
  • DescriptionEthrnet Swtch8port 10/100BaseTX Din-Rail
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
The 304TX, 306TX and 308TX are unmanaged industrial ethernet switches offering four, six or eight copper ports. Housed in rugged DIN-RAIL enclosures, they're designed for use in industrial data acquisition, control and Ethernet I/O applications.