Red Lion 306FX2-SC Ethrnet Swtch6prt 4 10/100BaseTX 2 100BaseFX Din-Rail
Representative image only.
Red Lion Mfr # 306FX2-SC – Item # EW-17782-95
$604.00USD / Each
sign in for your price.
SPECS & DESCRIPTION
Specifications & Description
  • Number of Ports6
  • Max Input Voltage (VDC)30
  • Min Input Voltage (VDC)10
  • Max Input Current (mA)250
  • DescriptionEthrnet Swtch6prt 4 10/100BaseTX 2 100BaseFX Din-Rail
More About this Item
The 302MC is a 10/100 BaseTX to 100BaseFX Industrial Media Converter. The 305FX and 306FX2 are unmanaged industrial ethernet switches with four copper and one or two fiber ports. Housed in a rugged DIN-RAIL enclosure, these devices ared esigned for use in industrial data acquisition, control and Ethernet I/O applications.