Red Lion 305FX-ST Ethrnet Swtch5port 4 10/100BaseTX 1 100 Basefx Din-Rail
Representative image only.
Red Lion Mfr # 305FX-ST – Item # EW-17793-03
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Number of Ports5
  • Max Input Voltage (VDC)30
  • Min Input Voltage (VDC)10
  • Max Input Current (mA)250
  • DescriptionEthrnet Swtch5port 4 10/100BaseTX 1 100 Basefx Din-Rail
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
The 302MC is a 10/100 BaseTX to 100BaseFX Industrial Media Converter. The 305FX and 306FX2 are unmanaged industrial ethernet switches with four copper and one or two fiber ports. Housed in a rugged DIN-RAIL enclosure, these devices ared esigned for use in industrial data acquisition, control and Ethernet I/O applications.