Reagents Calcium Chloride Anhydrous, 500g, Purified

Representative image only.
Reagents Mfr # C2162550-500A5 – Item # EW-78910-69
Manufactured using the highest quality raw materials in ISO 9001 facility
  • Reagents and solutions to meet your lab and research needs
  • 7 year warranty

$69.20USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradePurified
  • Linear FormulaCaCl2
  • ClassSalts
  • FormSolid
  • Weight (g)500
  • Formula Weight110.98
  • PackagingPoly Jar
  • CAS Number10043-52-4
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • DescriptionCalcium Chloride Anhydrous, 500g, Purified
  • Warranty7 year

Key Features

  • Reagents and solutions to meet your lab and research needs
  • 7 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat