RAMCO AA-002-01 Acid Spot'r Acid Detecting Spray, 16oz Trigger Spray
Representative image only.
Mfr # AA-002-01 – Item # EW-08779-62
Protect employees, equipment, and the environment
PRODUCT OPTIONS
$86.60USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestpH
  • DescriptionAcid Spot'r Acid Detecting Spray, 16oz Trigger Spray
More About this Item
ACID SPOT’R™ is a translucent yellow spray that immediately turns purple upon contact with even small drops of acid pH 2.6 or lower. This is a water soluble product used for the detection of accidental spills or leaks of acids. Use whenever an acid spill or leak occurs or is suspected. ACID SPOT’R™ will determine the perimeter of a spill and will confirm the isolation, repair, and neutralization of a leak. Use this product before coating a potentially contaminated surface to check for surface contamination.