Ralston Instruments QTHA-KIT4 Universal Fitting Kit; 20 Adapters/Hoses/Case
Representative image only.
Ralston Instruments Mfr # QTHA-KIT4 – Item # EW-68976-74
Adapters have a unique sealing system that makes them excellent for both high and low pressure calibration.
  • ABS case with foam cut-outs makes it easy to see if any adapters are missing
  • Adapters are available in kits or individually.
  • 1 year warranty
PRODUCT OPTIONS
$1,652.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Pressure5000 psi (345 bar)
  • Min Pressure0 psi (0 bar)
  • DescriptionUniversal Fitting Kit; 20 Adapters/Hoses/Case
  • Warranty1 Year
1 Year Warranty
Key Features
  • ABS case with foam cut-outs makes it easy to see if any adapters are missing
  • Adapters are available in kits or individually.
  • 1 year warranty
More About this Item
Kits take the guesswork out of selecting adapters by providing a selection of the most popular combinations of hoses and adapters necessary to connect to any pressure transmitter, gauge, hand pump, manifold or nitrogen source. Use the individual hoses and fittings to connect to any device you will be calibrating.
Item Includes
1/2" female NPT Quick-connect x male Quick-test, no check-valve, S.S. (QTHA-4FS0-QD)
1/2" female NPT x male Quick-test, no check-valve, brass (QTHA-4FB0)
1/2" male NPT x male Quick-test, no check-valve, brass (QTHA-4MB0)
1/2" tube fitting x male Quick-test, no check-valve, brass (QTHA-4TB0)
1/4" female NPT Quick-connect x male Quick-test, no check-valve, S.S. (QTHA-2FS0-QD)
1/4" female NPT x male Quick-test, no check-valve, brass (QTHA-2FB0)
1/4" male NPT QD (Quick-connect) Adapter, S.S. (QTHA-2MS0-QD)
1/4" male NPT x male Quick-test, no check-valve, brass (QTHA-2MB0)
1/4" tube fitting x male Quick-test, no check-valve, brass (QTHA-2TB0)
1/8" female NPT Quick-connect x male Quick-test, no check-valve, S.S. (QTHA-1FS0-QD)
1/8" female NPT x male Quick-test, no check-valve, brass (QTHA-1FB0)
1/8" male NPT QD (Quick-connect) Adapter, S.S. (QTHA-1MS0-QD)
1/8" male NPT x male Quick-test, no check-valve, brass (QTHA-1MB0)
3/8" female NPT x male Quick-test, no check-valve, brass (QTHA-3FB0)
3/8" male NPT x male Quick-test, no check-valve, brass (QTHA-3MB0)
3/8" tube fitting x male Quick-test, no check-valve, brass (QTHA-3TB0)
Honeywell 1/4"-28 UNF X male Quick-test, no check-valve, S.S. (QTHA-HNS0)
Male Quick-test run x male Quick-test run x male Quick-test branch tee, brass (QTHA-HSTE)
Male Quick-test x male Quick-test hose union, brass (QTHA-HSHS)
Quick-test 6900 psi hose, brass hose ends, 3 ft (92 cm) long (QTQT-HOS-3ft)
Quick-test 6900 psi hose, brass hose ends, 5 ft (1.52 m) long (QTQT-HOS-5ft)
Rosemount 5/16"-24 UNF x male Quick-test, no check-valve, S.S. (QTHA-FRS0)