PrimeLock® Installation Mandrel, minimum tube union, 3/4" fittings; 1/ea
Representative image only.
Item # EW-30491-24
Optimize performance of your PrimeLock® system
  • Choose from a wide range of replacement parts and accessories
PRODUCT OPTIONS
$96.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPFA
  • Connection TypeFlare Installation Mandrel
  • Compression Size / Tube Od (Port 1) (in)3/4
  • ColorNatural
  • DescriptionInstallation Mandrel, minimum tube union, 3/4" fittings; 1/ea
Key Features
  • Choose from a wide range of replacement parts and accessories
More About this Item
Installation tools require mandrel(s) and clamp insert(s) for operation. EPDM-lined clamp inserts eliminate surface marks on tubing following insert installation. Minimum tube union installations require the appropriately sized minimum tube union mandrel in addition to the standard installation mandrel. Directional sleeve mandrels prevent backwards installation of the insert, sizes 3/8" to 1-1/4".