Prepared media tube, EC broth with MUG—prepared tube, 20 pack
Representative image only.
Item # EW-14203-66
Save time and money with ready-made tubes
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
  • For detection of E. coli in food and water by fluorescence
  • Contains methylumbelliferyl glucuronide
  • Contains durham tube for gas detection
  • Tube size: 20 x 125 mm
  • 3 month warranty
PRODUCT OPTIONS
$99.50USD / Pkg of 20
SPECS & DESCRIPTION
Specifications & Description
  • Media Name (No.)E.C. w/ MUG
  • DescriptionPrepared media tube, EC broth with MUG—prepared tube, 20 pack
  • Warranty3 month
3 Month Warranty
Key Features
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
  • For detection of E. coli in food and water by fluorescence
  • Contains methylumbelliferyl glucuronide
  • Contains durham tube for gas detection
  • Tube size: 20 x 125 mm
  • 3 month warranty
More About this Item
Tubes are tested and standardized to ensure consistent and reliable performance. Each tube includes a certificate of compliance and quality control that shows lot numbers, expiration dates, and shelf-life guarantee. Tube and pack sizes vary.