Porta-Lab HP-2003 Portable Laboratory Hot Plate Butane
Representative image only.
Porta-Lab Mfr # HP-2003(1 UNIT) – Item # EW-36138-00
Gas-operated hot plate provides an efficient source of infrared heat
  • Solid-state push-button auto ignition
  • Heat output is fully adjustable
$66.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Gas TypeButane
  • Height (in)3
  • DescriptionPortable Laboratory Hot Plate Butane
Key Features
  • Solid-state push-button auto ignition
  • Heat output is fully adjustable
More About this Item
This compact ceramic heating burner features an infrared heat source that can be adjusted from ambient to 2012°F (1100°C). Gas operation makes it available for use without the need of an electrical outlet. Easily refill hot plate with standard butane fuel; order cartridges separately below. The optional beaker stand is made of high-quality chrome and supports a 1000-mL breaker. Fuel capacity is 40 gms (1 to 2 hrs/fill). Unit measures 3" H x 6" dia.