Porta-Lab EW-M1 Butane Fuel Canisters, 1.8 oz Cans; 12Cans/Case

Representative image only.
Porta-Lab Mfr # EW-M1 – Item # EW-36138-90
Enhance the performance of your Argos Technologies® burners
  • Premium fuel
  • Universal filling tube
  • Odorant added to detect leakage

PRODUCT OPTIONS

$86.50USD / Case of 1

SPECS & DESCRIPTION

Specifications & Description

  • Gas TypeButane
  • DescriptionButane Fuel Canisters, 1.8 oz Cans; 12Cans/Case

Key Features

  • Premium fuel
  • Universal filling tube
  • Odorant added to detect leakage

More About this Item

A variety of fuel adapters and holders to accommodate different burners and applications.

Canister Unit Size: 1.5 in x 6.5 in
Unit Fuel Capacity: 1.8 oz

04397, 04397-1, 04397-2, 04397 1, 04397 2, EW, GH, SI, AO

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat