Pomona Electronics 72923-0 Mini Test Probeblck With Sharp Spring Mounted Tip
Representative image only.
Pomona Electronics Mfr # 72923-0 – Item # EW-20009-68
Available for sale only in the United States and Canada.
  • 3 month warranty
$21.59USD / Each
$23.99USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionMini Test Probeblck With Sharp Spring Mounted Tip
3 Month Warranty
Key Features
  • 3 month warranty
More About this Item
SMD test probe with sharp, spring mounted tip and 2mm jack. Provides ultra-fine control for probing high density SMD circuits. Rating: 600V, CAT III, 1A.