Cole-Parmer Essentials Sterile Polystyrene Plastic Lab Forceps, Serrated Blunt Tips, 12.2 cm L (4.8”); 100/PK
Representative image only.
Cole-Parmer Essentials Item # EW-06443-20
Corrugated handles provide a secure finger grip for better control
  • Disposable - reduces cross contamination
  • Serrated tips for accurate sample grip
  • Sterile individually bagged and non-sterile options
  • Individually wrapped
PRODUCT OPTIONS
$173.00USD / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • Overall Length (in)4 51/64
  • Overall Length (cm)12.2
  • MaterialPolystyrene
  • ColorBlue
  • SterileYes
  • StyleSerrated Blunt Tips
  • DescriptionSterile Polystyrene Plastic Lab Forceps, Serrated Blunt Tips, 12.2 cm L (4.8”); 100/PK
Key Features
  • Disposable - reduces cross contamination
  • Serrated tips for accurate sample grip
  • Sterile individually bagged and non-sterile options
  • Individually wrapped
More About this Item
Reduce cross contamination with these disposable forceps. Choose from individually bagged, sterile (reinforced nylon or polystyrene) or non-sterile (reinforced nylon) lab forceps to best suit your needs. All plastic forceps have corrugated handles to provide you with a secure finger grip. Serrated tips allow you to accurately grip the sample. Use serrated pointed tips for handling smaller objects or serrated blunt tips for general use. Nylon forceps are autoclavable.
Certificate of Analysis
For a sample Certificate of Analysis, please go to link below under RESOURCES.
If a Lot specific Certificate of Analysis is required, please call 1-800-323-4340 for assistance.