PFA In-Line Filter Holder, 47-mm, 1/4" OD x 1/4" OD Tubing Connections

Representative image only.
Item # EW-06103-13
Nonwetting and nonsticking filter holder
  • Temperature range is –320 to 500°F (–196 to 260°C)
  • Withstands up to 65 psi
  • 3 month warranty

PRODUCT OPTIONS

$320.50USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Filter Diameter (mm)47
  • Body MaterialPFA
  • DescriptionPFA In-Line Filter Holder, 47-mm, 1/4\" OD x 1/4\" OD Tubing Connections
  • Warranty3 month
3 Month Warranty

Key Features

  • Temperature range is –320 to 500°F (–196 to 260°C)
  • Withstands up to 65 psi
  • 3 month warranty

More About this Item

Suitable for organic solvents, acids, alkalies, or high-purity filtration. Change filter without disconnecting tubing with Tefzel® ETFE clamp nut. Prefiltering reservoir creates fluid turbulence to slow membrane clogging. PFA support grid snaps out easily for cleaning. Ferruled nuts accept 1/4'' OD PTFE or PFA tubing. Order filter membranes separately.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat