Pentair WPX5BB97P Fibrillated polypropylene string-wound cartridge, 5 µm, 10" length
Representative image only.
Pentair Mfr # WPX5BB97P – Item # EW-29802-40
String-wound pattern provides greater surface area for lower pressure drop and higher flow
PRODUCT OPTIONS
$37.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaPolypropylene
  • Micron Rating5
  • Filter Length (in)10
  • DescriptionFibrillated polypropylene string-wound cartridge, 5 µm, 10" length
More About this Item
Polypropylene string is resistant to chemical and bacterial attack. Suitable to 165°F (74 °C).
Fibrillated cartridges are non-leaching and offer minimal fiber migration with a higher pressure drop.