Pentair WPX100BB20P Fibrillated polypropylene string-wound cartridge, 100 µm, 20" length
Representative image only.
Pentair Mfr # WPX100BB20P – Item # EW-29802-45
String-wound pattern provides greater surface area for lower pressure drop and higher flow
PRODUCT OPTIONS
$65.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaPolypropylene
  • Micron Rating100
  • Filter Length (in)20
  • DescriptionFibrillated polypropylene string-wound cartridge, 100 µm, 20" length
More About this Item
Polypropylene string is resistant to chemical and bacterial attack. Suitable to 165°F (74 °C).
Fibrillated cartridges are non-leaching and offer minimal fiber migration with a higher pressure drop.