Pentair 355215-43 Fibrous polypropylene string-wound cartridge, 10 µm, 10" length
Representative image only.
Pentair Mfr # 355215-43 – Item # EW-29802-35
String-wound pattern provides greater surface area for lower pressure drop and higher flow
PRODUCT OPTIONS
$32.75USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaPolypropylene
  • Micron Rating10
  • Filter Length (in)10
  • DescriptionFibrous polypropylene string-wound cartridge, 10 µm, 10" length
More About this Item
Polypropylene string is resistant to chemical and bacterial attack. Suitable to 165°F (74 °C).
Fibrous polypropylene cartridges have a lower pressure drop than fibrillated cartridges but use an antistatic agent that may cause foaming in some applications.