Pentair 155263-03 Double open end iron reduction cartridge; 20"L
Representative image only.
Pentair Mfr # 155263-03 – Item # EW-29830-62
$164.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaPermanganese
  • Flow Rate6 GPM
  • Filter Length (in)20
  • CapacityUp to 3ppm dissolved iron
  • DescriptionDouble open end iron reduction cartridge; 20"L
More About this Item
Iron Reduction Cartridge provides an easy yet effective way to reduce iron in water. Reduces the metallic taste of water and helps eliminate stains on plumbing fixtures. Ideal for drinking water.