PCRmax RNA Reston ebolavirus, with mastermix
Representative image only.
PCRmax Item # EW-93944-31
Crucial tool for fast and accurate detection of dangerous human viral pathogens
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
SPECS & DESCRIPTION
Specifications & Description
  • Test TypeRNA
  • Number of Tests Included150
  • Mastermix IncludedYes
  • DescriptionReston ebolavirus, with mastermix
Includes
Onestep mastermix (RNA transcriptase), lyophilized mastermix, lyophilized ROX, resuspension buffer, lyophilized primer and probe mix, positive control for standard curve, internal extraction control RNA, endogenous control, and RNAse/DNAse-free water.
Key Features
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
More About this Item
These detection kits utilize the sensitivity and speed of qPCR to get the most accurate data as quickly as possible. Each kit is specifically designed by our bioinformatics team to ensure the broadest possible detection profile and detection of all relevant strains and subtypes. All test kits are validated in house on multiple qPCR platforms to ensure cross platform functionality. Kits come with sufficient controls to reduce the chances of making the wrong call. Kits can also by generated specifically for your test of interest. If the kit is not in our database, we can easily make the kit you wish if sequence data is available. Kits conform to the latest safety directives including European CE requirements.
Order a complete kit with mastermix or a kit without mastermix and use your own mastermix.