Parker Hannifin Inserts for soft tubing, white PP, 5/16" OD x 3/16" ID; 10/Pk
Representative image only.
Parker Hannifin Mfr # P-P-5 – Item # EW-06391-20
Allows compression fittings to be used with soft tubing
  • Secure your flexible, soft tubing
PRODUCT OPTIONS
$8.40USD / Pkg of 10
BUY MORE & SAVE
  • 1-9$8.40USD / Pkg of 10
  • 10-49$7.56USD / Pkg of 10
  • 50-99$7.14USD / Pkg of 10
  • 100+$6.72USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolypropylene
  • Sterilization MethodEthylene oxide (ETO) only
  • Connection TypeCompression
  • Barb Size / Tubing Id (Port 2) (in)3/16
  • Compression Size / Tube Od (Port 1) (in)5/16
  • ColorWhite
  • DescriptionInserts for soft tubing, white PP, 5/16" OD x 3/16" ID; 10/Pk
Key Features
  • Secure your flexible, soft tubing
More About this Item
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Inserts fit in the end of the soft tubing to provide the rigidity required for adequate compression and a proper seal.