Dynalon Clear acrylic dispenser and cutter for 4" wide Parafilm rolls

Representative image only.
Dynalon Mfr # 176205 – Item # EW-06720-34
Cuts desired Parafilm sheets without scissors
  • Securely holds 4" wide parafilm rolls
  • Attached cutting blade

$241.00USD / Each
Often bought with...

SPECS & DESCRIPTION

Specifications & Description

  • ColorClear
  • MaterialAcrylic
  • Width (in)4
  • Width (cm)10.2
  • DescriptionClear Acrylic Dispenser and Cutter for 4\" Wide Film

Key Features

  • Securely holds 4" wide parafilm rolls
  • Attached cutting blade

More About this Item

Acrylic dispenser provides dust-free storage and serrated cutting surface for easy dispensing of 4” Parafilm® rolls.

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat