Panasonic NE-1054 Programmable Countertop Microwave Oven, 1000 Watts, 0.8 Cu. Ft., Stainless Steel; 120V, 60 Hz

Representative image only.
Panasonic Mfr # B005GSPVHS – Item # EW-24406-39
Economical microwave ovens for general laboratories or employee break rooms
  • Programmable for a wide variety of heating tasks
  • Sensor automatically adjust time and power for greater control of heating

PRODUCT OPTIONS

$1,300.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (cu ft)0.8
  • Exterior Width (in)20 1/8
  • Exterior Height (in)12
  • Exterior Depth (in)15 3/4
  • Control TypeTouch control keypad
  • DisplayDigital
  • Power (VAC)120
  • Power (Hz)60
  • Power (amps)13.4
  • Power (watts)1000
  • ConstructionStainless Steel
  • DescriptionProgrammable Countertop Microwave Oven, 1000 Watts, 0.8 Cu. Ft., Stainless Steel; 120V, 60 Hz

Key Features

  • Programmable for a wide variety of heating tasks
  • Sensor automatically adjust time and power for greater control of heating

More About this Item

These economical microwaves feature 10 programmable memory pads which make quick work of heating your items. Ideal for light operations, these microwave ovens can help you heat and serve small items with ease. The stainless-steel door can stand up to everyday use and is easily cleaned. Six power levels enable you to heat up a variety of items

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat