Oakton waterproof DO meter replacement probe with 25-ft cable
Representative image only.
Oakton Item # EW-35640-52
Optimize oxygen analysis
  • For a variety of Oakton® models
  • 1 year warranty
PRODUCT OPTIONS
$809.00USD / Each
DO 6+ Dissolved Oxygen Meters
Oakton® DO 6+ Dissolved Oxygen Meters
ITEM(S) ON SALE
$535.00 - $986.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithOakton 600 series DO meters
  • Length (in)300
  • Descriptionwaterproof DO meter replacement probe with 25-ft cable
  • Warranty1 year
1 Year Warranty
Key Features
  • For a variety of Oakton® models
  • 1 year warranty
More About this Item
Probes for dissolved oxygen and biological oxygen demand for Oakton DO/BOD oxygen meters.